Orders LinkContactUsLinkComponent
Test kits, drop count tests
Test kits, drop count tests
Chloride, model CD-51, 500 - 10000 mg/l Cl-
5000 - 100000 mg/l Cl-, 1 KIT
Catalog # HACH208601
Supplier:   Hach
Test kits, drop count tests
Catalog # HACH208601
Supplier:  Hach
CAS Number:  
Test kits, drop count tests
Test kits, drop count tests
Chloride, model CD-51, 500 - 10000 mg/l Cl-
5000 - 100000 mg/l Cl-, 1 KIT
CAS Number:  
Price
435.00 SFr.
Log in for availability
1 * 1 KIT
Product Details & Documents
The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.
  • Portable - ideal for use in field or in lab
  • Used as a quick reference or for confirmatory test with colour indicator
  • Reagents supplied with the kit
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    Chloride, model CD-51
  • Range:
    500 - 10000 mg/l Cl-<br />5000 - 100000 mg/l Cl-