Paraformaldehyde, powder, TECHNICAL
Plastic bottle for solids, 1 kg
Supplier: VWR Chemicals
Paraformaldehyde, powder, TECHNICAL
Catalog # 28794.295
Supplier: VWR Chemicals
CAS Number: 30525-89-4
Jump to:
Product Details & Documents
Synonyms: Polyoxymethylene
- CAS Number:30525-89-4
- Formula:(CH₂O)n
- MDL Number:MFCD00133991
- Merck Index:13,07096
- Boiling Pt:135…136 °C (13 Torr)
- Melting Pt:120…170 °C
- Density:0.88 g/cm³ (25 °C)
- Flash Pt:70 °C (closed cup)
- Storage Temperature:Ambient
- UN:2213
- ADR:4.1,III
Specifications
Specification Test Results
- Identification:Passes test
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



