Sodium hydroxide 35% in aqueous solution, TECHNICAL
Plastic drum, 20 L
Supplier: VWR Chemicals
Sodium hydroxide 35% in aqueous solution, TECHNICAL
Catalog # 28221.445
Supplier: VWR Chemicals
CAS Number: 1310-73-2
Jump to:
Product Details & Documents
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Boiling Pt:119…135 °C (1013 hPa)
- Melting Pt:9…10 °C
- Density:1,53 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-185-5
- UN:1824
- ADR:8,II
Documents
Certificates
Search by Lot Number1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 200L Plastic drums for liquids UN (ZPACK13950512) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
P20_VWRC28221.365_SPEC_EN.pdf P20_VWRC28221.445_SPEC_EN.pdf spec_VWRC28221.296 spec_VWRC28221.296 spec_VWRC28221.365
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Specifications
Specification Test Results
- Assay:Min. 34 %
- Heavy metals (as Pb):Max. 50 ppm
- Fe (Iron):Max. 50 ppm
Specifications
- Pack type:Plastic drum
- Pk:20 L



