Sodium hydroxide 30% in aqueous solution, TECHNICAL
Plastic drum, 20 L
Supplier: VWR Chemicals
Sodium hydroxide 30% in aqueous solution, TECHNICAL
Catalog # 28217.441
Supplier: VWR Chemicals
CAS Number: 1310-73-2
Jump to:
Product Details & Documents
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Boiling Pt:119…135 °C (1013 hPa)
- Melting Pt:9…10 °C
- Density:1,53 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-185-5
- UN:1824
- ADR:8,II
Dokumente
Certificates
Search by Lot Number1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 200L Plastic drums for liquids UN (ZPACK13950512) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Verpackungsinformationen
P20_VWRC28217.292_SPEC_EN.pdf P20_VWRC28217.361_SPEC_EN.pdf P20_VWRC28217.441_SPEC_EN.pdf P20_VWRC28217.554_SPEC_EN.pdf spec_VWRC28217.292
Produktspezifikation
More
SDS_EN_UK_VWRC28217.pdf sds_VWRC28217
Sicherheitsdatenblatt (SDS)
VWR Chemicals Big Packs and Bulks
Produktliteratur
Specifications
Specification Test Results
- Assay:Min. 29 %
- Heavy metals (as Pb):Max. 50 ppm
Specifications
- Pack type:Plastic drum
- Pk:20 L



