Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Methylcyclohexane ≥99%, TECHNICAL

Metal drum, 25 L
  25505.468
 
 
 
 
Methylcyclohexane ≥99%, TECHNICAL
  25505.468
 :  108-87-2
Methylcyclohexane ≥99%, TECHNICAL
Methylcyclohexane ≥99%, TECHNICAL

Metal drum, 25 L

 :  108-87-2
 
SFr. 679.00
   
1 * 25 L
 
  •  :108-87-2
  •  :C₆H₁₁CH₃
  •  :MFCD00001497
  •  :98.19 g/mol
  •  :13,06074
  •  :101 °C (1013 hPa)
  •  :–126 °C
  •  :0.7693 g/cm³ (20 °C)
  •  :–4 °C
  •  :Ambient
  •  :203-624-3
  •  :2296
  •  :3,II
 

 

  • Assay:
    Min. 99 %
  • Identification:
    Passes test
  • Water:
    Max. 50 ppm

 

  • Pack type:
    Metal drum
  • Pk:
    25 L

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.