ContactUsLinkComponent
GASTUBE METHYL CHLOROFORM 100-2000PPM 1 * 10 items
Catalog # GAST135
Supplier:  GASTEC
GASTUBE METHYL CHLOROFORM 100-2000PPM 1 * 10 items
Catalog # GAST135
Supplier:  GASTEC
Frequently Bought Together