Orders LinkContactUsLinkComponent
Instrument boxes
Instrument boxes
Red, 100×215×50 mm
Catalog # AGEN8036R
Supplier:   Agencinox
Instrument boxes
Catalog # AGEN8036R
Supplier:  Agencinox
Instrument boxes
Instrument boxes
Red, 100×215×50 mm
Price
SFr. 72.40
Log in for availability
1 * 1 ST
Product Details & Documents
Aluminium, anodised.
  • Smooth surfaces for fast, easy cleaning
  • Coloured
  • Good chemical resistance, very good thermal resistance
Specifications
  • Colour:
    Red
  • Seq:
    08
  • W×D×H:
    100×215×50 mm