Spatulas
Spatula, lab type, double (C), 200 mm
Catalog # 231-0002
Supplier: Agencinox
Jump to:
Product Details & Documents
Stainless steel ¹⁸/₁₀, anti-magnetic, electro-polishing. These universal spatulas are suitable for many applications.
- Excellent chemical and thermal resistance
- Autoclavable
Documents
Specifications
- Description:Spatula, lab type, double (C)
- Length:200 mm