Spatulas
Spatula, semi-micro (D), 140 mm
Catalog # 231-0001
Supplier: Agencinox
Jump to:
Product Details & Documents
Stainless steel ¹⁸/₁₀, anti-magnetic, electro-polishing. These universal spatulas are suitable for many applications.
- Excellent chemical and thermal resistance
- Autoclavable
Documents
Specifications
- Description:Spatula, semi-micro (D)
- Length:140 mm