Orders LinkContactUsLinkComponent
Spatulas
Spatula, spear-shaped (G), 140 mm
Catalog # 231-0000
Supplier:   Agencinox
Spatulas
Catalog # 231-0000
Supplier:  Agencinox
Spatulas
Spatulas
Spatula, spear-shaped (G), 140 mm
Price
SFr. 64.20
Log in for availability
1 * 5 ST
Product Details & Documents
Stainless steel ¹⁸/₁₀, anti-magnetic, electro-polishing. These universal spatulas are suitable for many applications.
  • Excellent chemical and thermal resistance
  • Autoclavable
Specifications
  • Description:
    Spatula, spear-shaped (G)
  • Length:
    140 mm