Screw caps, bakelite
with rubber liner, 30 mm, Black
Catalog # 215-2678
Supplier: GRAVIS
Jump to:
Product Details & Documents
Specifications
- Thread:30 mm
- Colour:Black
- Description:with rubber liner
- Seq:12