Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Roller bottles, WHEATON®

Roller bottles, WHEATON®

Roller bottles, with 38 mm deep skirted, rubber lined black phenolic cap, 3450 ml, 1320 cm², 110×480 mm, 4/case
Catalog # 734-3019
Supplier:   DWK Life Sciences
Roller bottles, WHEATON®
Catalog # 734-3019
Supplier:  DWK Life Sciences
Roller bottles, WHEATON®
Roller bottles, WHEATON®

Roller bottles, with 38 mm deep skirted, rubber lined black phenolic cap, 3450 ml, 1320 cm², 110×480 mm, 4/case

Price
SFr. 881.00
Log in for availability
1 * 4 ST
Product Details & Documents

Rollers bottles are specifically designed for adherent cell roller culture

  • Bottles withstands repeated wet or dry sterilization cycles and are reusuable
  • These culture vessels are made to exacting tolerances in order to provide higher cell yields
  • Made of USP Type I borosilicate glass

Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904

Specifications
  • Growth area:
    1320 cm²
  • Packed:
    4/case
  • Capacity:
    3450 ml
  • Description:
    Roller bottles, with 38 mm deep skirted, rubber lined black phenolic cap
  • Ø ext.×H:
    110×480 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.