Stopcocks, straight through, with tip
Glass
Catalog # COWI016.9212.1
Supplier: COWIE
Jump to:
Product Details & Documents
Made of PTFE.
- Available with PP or glass tip
- Very good chemical resistance
- Very good temperature resistance
Specifications
- Description:Glass