Counting chambers, Thoma
Counting chamber with double ruling, Thoma, 0,10 mm
631-1151
For counting thrombocytes and erythrocytes, the grid system corresponds to the large square in the middle of the Neubauer chamber.
- Subdivision doubles the analysis options
- Bright-line division increases contrast of the net system to make work much easier
- Spring clips guarantee the exact positioning of the cover slips
- For blood capillaries
- Each small square has a surface of 0,0025 mm²
- Depth of chamber:0,10 mm
- Description:Counting chamber with double ruling, Thoma



