Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Linen testers

Linen testers

14,6 mm, 15×15 mm
Catalog # 630-8326
Supplier:   ESCHENBACH OPTIK
Linen testers
Catalog # 630-8326
Supplier:  ESCHENBACH OPTIK
Linen testers
Linen testers

14,6 mm, 15×15 mm

Price
SFr. 78.40
Log in for availability
1 * 1 ST
Product Details & Documents

Satin matt chrome plated brass frame with silicate glass lens.

Specifications
  • Ø:
    14,6 mm
  • Magnification:
    10×
  • W×D:
    15×15 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.