Folding precision magnifiers
Folding magnifier, achromatic, 17 mm
Catalog # 630-8222
Supplier: ESCHENBACH OPTIK
Jump to:
Product Details & Documents
Swivelling black metallic magnifier with loop, lens frame made from black painted metal.
- Protective case can serve as a handle
- Provide distortion-free images
- Chrome plated metal
Specifications
- Ø:17 mm
- Magnification:20×
- Description:Folding magnifier, achromatic
- Seq:004