Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Storage Boxes

Storage Boxes

5×10, 140×70×36 mm, 12 mm
Catalog # 391-6191
Marketsource
Storage Boxes
Catalog # 391-6191
Marketsource
Storage Boxes
Storage Boxes

5×10, 140×70×36 mm, 12 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Boxes, made of PS, clear, with loose inserts.

Specifications
  • For tubes:
    12 mm
  • No. of Holes:
    5×10
  • Dimensions:
    140×70×36 mm
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.