Scalpels, ceramic
36, 145 mm, 25 mm
233-5094
Particularly high quality and durable hard ceramic material for long lasting, excellent sharpness. No metal parts, thus also suitable for trace analysis. Super-lightweight, sturdy, ergonomic handle for safe and easy handling.
- Anti-magnetic, anti-static, corrosion-free, dishwasher safe, chemically resistant to aggressive media
- Extremely heat resistant, low thermal conductivity
- Regrinding is not necessary
- Type:36
- Length:145 mm
- Seq:04
- Blade:25 mm



