Orders LinkContactUsLinkComponent
Forceps, ESD-safe
Curved, hook-shaped fine tips, NK6, Black, 120 mm
  232-2171
Forceps, ESD-safe
  232-2171
Forceps, ESD-safe
Forceps, ESD-safe
Curved, hook-shaped fine tips, NK6, Black, 120 mm
SFr. 24.30
1 * 1 ST
Conductive Vestamid with 25% fibre glass reinforcement, black. Precise, robust and virtually unbreakable forceps. Very precise; when pressure is applied during work, the tips don’t open.
  • Electrically conductive
  • Very high chemical resistance to acids and bases
  • Heat resistant up to 175 °C
DIN 53482
 : DIN 53482
  • Type:
    NK6
  • Colour:
    Black
  • Description:
    Curved, hook-shaped fine tips
  • Length:
    120 mm