Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Scoops

Scoops

1600 ml, 400 mm
Catalog # 231-0308
Supplier:   SCHROEDER WILESCO
Scoops
Catalog # 231-0308
Supplier:  SCHROEDER WILESCO
Scoops
Scoops

1600 ml, 400 mm

Price
22.40 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents

Aluminium, smooth. For collecting or measuring powders, granulates or pastes.

  • Good chemical resistance
  • Excellent thermal resistance
  • Light, robust and easy to use
Specifications
  • Capacity:
    1600 ml
  • Length:
    400 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.