Dropping bottles, Lucas
100 ml
Katalognr. 215-9106
Anbieter: MUELLER KREMPEL
Direkt zu
Product Details & Documents
Amber glass, without closure.
- Good chemical and physical properties
- Use in applications that do not involve high thermal stresses
- Please order screw caps and drop dispensers separately
Brown coloured to protect light-sensitive materials.
Technische Daten
- Capacity:100 ml
- Seq:09