Orders LinkContactUsLinkComponent
Dropping bottles, Lucas
Dropping bottles, Lucas
20 ml
Catalog # 215-9103
Supplier:   MUELLER KREMPEL
Dropping bottles, Lucas
Catalog # 215-9103
Supplier:  MUELLER KREMPEL
Dropping bottles, Lucas
Dropping bottles, Lucas
20 ml
Price
79.40 SFr.
Log in for availability
1 * 180 ST
Product Details & Documents
Amber glass, without closure.
  • Good chemical and physical properties
  • Use in applications that do not involve high thermal stresses
  • Please order screw caps and drop dispensers separately
Brown coloured to protect light-sensitive materials.
Specifications
  • Capacity:
    20 ml
  • Seq:
    05