Orders LinkContactUsLinkComponent
Replacement caps
Replacement caps
SVL 18
Catalog # 215-2043
Supplier:   QVF LABORTECHNIK
Replacement caps
Catalog # 215-2043
Supplier:  QVF LABORTECHNIK
Replacement caps
SVL 18
Price
SFr. 7.90
Log in for availability
1 * 1 ST
Product Details & Documents
Pyrex®, phenoplast, black.
  • Excellent chemical resistance, excellent temperature resistance
  • Screw caps with PTFE-coated seal
  • Replacement seals are available as an option
Replacement closures for reagent and sample glasses as well as bottles with SVL threads.
Specifications
  • Thread:
    SVL 18