Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Bottle Carriers

Silver, 6×1000 ml bottles, 110×110 mm
  215-0164
Bottle Carriers
  215-0164
Bottle Carriers
Bottle Carriers

Silver, 6×1000 ml bottles, 110×110 mm

 
SFr. 323.00
   
1 * 1 ST
 

Stainless steel or steel, coated.

  • Four non slip feet
  • Sturdy carrying handle
  • Good chemical and temperature resistance

These bottle carriers are suitable for transporting and storing bottles.

Colour: Silver/white

 

 
  • For:
    6×1000 ml bottles
  • Colour:
    Silver
  • Seq:
    05
  • Compartment size:
    110×110 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.