Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Racks for test tubes | Avantor

Racks for test tubes

24 (2×12), Stainless steel, 250×50×68 mm
Catalog # 212-6470
Supplier:   HUPFER METALLWERKE
Racks for test tubes
Catalog # 212-6470
Supplier:  HUPFER METALLWERKE
Racks for test tubes
Racks for test tubes

24 (2×12), Stainless steel, 250×50×68 mm

Price
SFr. 168.00
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel, rust-proof, electrolytically or steel polished, polyamide coated.

  • Excellent chemical resistance
  • Excellent thermal resistance (polyamide coating resistant up to 80 °C)
  • Robust, stable construction and easy to clean

Wire racks with second level tier, 21 mm high. Total height 68 mm. Hole size 18×18 mm.

Specifications
  • No. of Holes:
    24 (2×12)
  • Material:
    Stainless steel
  • Seq:
    3
  • W×D×H:
    250×50×68 mm
Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..