Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Racks for test tubes

12 (2×6), Stainless steel, 130×50×68 mm
  212-6455
Racks for test tubes
  212-6455
Racks for test tubes
Racks for test tubes

12 (2×6), Stainless steel, 130×50×68 mm

 
SFr. 109.00
   
1 * 1 ST
 

Stainless steel, rust-proof, electrolytically or steel polished, polyamide coated.

  • Excellent chemical resistance
  • Excellent thermal resistance (polyamide coating resistant up to 80 °C)
  • Robust, stable construction and easy to clean

Wire racks with second level tier, 21 mm high. Total height 68 mm. Hole size 18×18 mm.

 
  • No. of Holes:
    12 (2×6)
  • Material:
    Stainless steel
  • Seq:
    1
  • W×D×H:
    130×50×68 mm