Reinstwassersysteme Ultra Clear plus, Leistung: 1,2 l/min, TOC-Wert: < 10 ppb, Endtoxingehalt: < 0,02 EU/ml 1 * 1 ST
Catalog # 172-6402
Supplier: EVOQUA WATER TECHNOLOGIES
Reinstwassersysteme Ultra Clear plus, Leistung: 1,2 l/min, TOC-Wert: < 10 ppb, Endtoxingehalt: < 0,02 EU/ml 1 * 1 ST
Catalog # 172-6402
Supplier: EVOQUA WATER TECHNOLOGIES
Frequently Bought Together
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection