Microplates, 96-well with glass inserts
Microplate, PP, 96-well with fixed glass inserts, chemically inert, ~1200 µl
391-1970
- Description:Microplate, PP, 96-well with fixed glass inserts, chemically inert
- Well volume:~1200 µl