Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Separating funnels, Squibb

Separating funnels, Squibb

1000 ml, 29/32 NS, 20 ml
Catalog # 527-1994
Supplier:   Lenz Laborglas
Separating funnels, Squibb
Catalog # 527-1994
Supplier:  Lenz Laborglas
Separating funnels, Squibb
Separating funnels, Squibb

1000 ml, 29/32 NS, 20 ml

Price
SFr. 169.00
Log in for availability
1 * 1 ST
Product Details & Documents

DURAN®, borosilicate glass, with NS-PE stopper.

With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. With or without graduations.

Documents

Specifications
  • Socket size:
    29/32 NS
  • Capacity:
    1000 ml
  • Division:
    20 ml