Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Conical separating funnels

Conical separating funnels

2000 ml, 29/32 NS, 50 ml
Catalog # 527-1206
Supplier:   Lenz Laborglas
Conical separating funnels
Catalog # 527-1206
Supplier:  Lenz Laborglas
Conical separating funnels
Conical separating funnels

2000 ml, 29/32 NS, 50 ml

Price
SFr. 285.00
Log in for availability
1 * 1 ST
Product Details & Documents

DURAN®, borosilicate glass, with NS-PE stopper.

With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Both with or without graduations.

Documents

Specifications
  • Socket size:
    29/32 NS
  • Capacity:
    2000 ml
  • Division:
    50 ml