Orders LinkContactUsLinkComponent
Conical separating funnels
Conical separating funnels
500 ml, 29/32 NS, -
Catalog # 527-1139
Supplier:   Lenz Laborglas
Conical separating funnels
Catalog # 527-1139
Supplier:  Lenz Laborglas
Conical separating funnels
Conical separating funnels
500 ml, 29/32 NS, -
Price
SFr. 141.00
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass, with NS-PE stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Both with or without graduations.
Documents
Specifications
  • Socket size:
    29/32 NS
  • Capacity:
    500 ml
  • Division:
    -