Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Conical separating funnels | Avantor

Conical separating funnels

1000 ml, 29/32 NS, 20 ml
Catalog # 527-1225
Supplier:   Lenz Laborglas
Conical separating funnels
Catalog # 527-1225
Supplier:  Lenz Laborglas
Conical separating funnels
Conical separating funnels

1000 ml, 29/32 NS, 20 ml

Price
SFr. 178.00
Log in for availability
1 * 1 ST
Product Details & Documents

DURAN®, borosilicate glass, with NS-PE stopper.

With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Both with or without graduations.

Documents

Specifications
  • Socket size:
    29/32 NS
  • Capacity:
    1000 ml
  • Division:
    20 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.