Conical separating funnels
500 ml, 29/32 NS, 10 ml
Catalog # 527-1214
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN®, borosilicate glass, with NS-PE stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Both with or without graduations.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
LENZ04000073_TDS_EN
Datasheet
Specifications
- Socket size:29/32 NS
- Capacity:500 ml
- Division:10 ml



