Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Conical separating funnels

100 ml, 19/26 NS, 5 ml
Catalog # 527-1222
Supplier:   Lenz Laborglas
Conical separating funnels
Catalog # 527-1222
Supplier:  Lenz Laborglas
Conical separating funnels
Conical separating funnels

100 ml, 19/26 NS, 5 ml

Price
SFr. 92.00
Log in for availability
1 * 1 ST
Product Details & Documents

DURAN®, borosilicate glass, with NS-PE stopper.

With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Both with or without graduations.

Specifications
  • Socket size:
    19/26 NS
  • Capacity:
    100 ml
  • Division:
    5 ml