Stopcocks, one-way
4,0 mm, 18,8 NS
Catalog # 205-2223
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN®, borosilicate glass, with solid glass or PTFE key.
- Replaceable NS keys
- Screw thread retaining device
- Leakage rate compliant with DIN 12540-1
Straight bore.
* with capillary side arms
** with rubber retaining device.
Certifications: DIN 12554.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Hole Ø:4,0 mm
- Seq:07
- Spigot:18,8 NS