Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Attachable screw thread tubes

Attachable screw thread tubes

GL 25, 22 mm, 100 mm
Catalog # 201-1606
Supplier:   Lenz Laborglas
Attachable screw thread tubes
Catalog # 201-1606
Supplier:  Lenz Laborglas
Attachable screw thread tubes
Attachable screw thread tubes

GL 25, 22 mm, 100 mm

Price
SFr. 6.30
Log in for availability
1 * 1 ST
Product Details & Documents

DURAN®, borosilicate glass, clear.

  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

DIN screw thread GL.

Documents

Specifications
  • Thread:
    GL 25
  • Tube ext.Ø:
    22 mm
  • Seq:
    03
  • Total length:
    100 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.