Orders LinkContactUsLinkComponent
Stopcocks for separating funnels, DURAN®
12,5 NS
  SCOT286433401
Stopcocks for separating funnels, DURAN®
  SCOT286433401
Stopcocks for separating funnels, DURAN®
12,5 NS
SFr. 810.00
1 * 25 ST
DURAN®, borosilicate glass, with polished short funnel, without key.
  • Very good chemical resistance
  • High temperature resistance
For glass manipulators.
  • Spigot:
    12,5 NS