Orders LinkContactUsLinkComponent
Stopcocks for separating funnels, DURAN®
Stopcocks for separating funnels, DURAN®
18,8 NS
Catalog # SCOT286430601
Supplier:   DWK Life Sciences
Stopcocks for separating funnels, DURAN®
Catalog # SCOT286430601
Supplier:  DWK Life Sciences
Stopcocks for separating funnels, DURAN®
18,8 NS
Price
SFr. 1’220.00
Log in for availability
1 * 25 ST
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, grounded and polished, without key.
  • Very good chemical resistance
  • Very good temperature resistance
For glass manipulators.
Specifications
  • Spigot:
    18,8 NS