Orders LinkContactUsLinkComponent
Stopcocks for separating funnels, DURAN®
Stopcocks for separating funnels, DURAN®
12,5 NS
Catalog # SCOT286426209
Supplier:   DWK Life Sciences
Stopcocks for separating funnels, DURAN®
Catalog # SCOT286426209
Supplier:  DWK Life Sciences
Stopcocks for separating funnels, DURAN®
12,5 NS
Price
SFr. 459.00
Log in for availability
1 * 25 ST
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, grounded, without key.
  • Very good chemical resistance
  • Very good temperature resistance
For glass manipulators.
Specifications
  • Spigot:
    12,5 NS