Filter discs, DURAN®
1, 90 mm, 6 mm
SCOT251590105
Sintered glass filters have neutral properties with unfused edge and untreated surfaces. The hard sintering process makes the material resilient.
- Available in intermediate sizes, alternative dimensions, alternate porosities and bi-planar design
Made from clear DURAN® borosilicate glass 3,3.
- Ø:90 mm
- Porosity:1
- Seq:32
- Thickness:6 mm
- Cat. No.:SCOT251590105



