Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Filter discs, DURAN®

4, 10 mm, 3 mm
Catalog # SCOT251510406
Supplier:   DWK Life Sciences
Filter discs, DURAN®
Catalog # SCOT251510406
Supplier:  DWK Life Sciences
Filter discs, DURAN®
Filter discs, DURAN®

4, 10 mm, 3 mm

Price
SFr. 72.20
Log in for availability
1 * 10 ST
Product Details & Documents

Sintered glass filters have neutral properties with unfused edge and untreated surfaces. The hard sintering process makes the material resilient.

  • Available in intermediate sizes, alternative dimensions, alternate porosities and bi-planar design

Made from clear DURAN® borosilicate glass 3,3.

Specifications
  • Ø:
    10 mm
  • Porosity:
    4
  • Seq:
    69
  • Thickness:
    3 mm