Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Burette stopcocks, lateral

Burette stopcocks, lateral

Spare PTFE key for 25 ml and 50 ml burette capacity
Catalog # BRND81248
Supplier:   Brand
Burette stopcocks, lateral
Catalog # BRND81248
Supplier:  Brand
Burette stopcocks, lateral
Burette stopcocks, lateral

Spare PTFE key for 25 ml and 50 ml burette capacity

Price
SFr. 28.10
Log in for availability
1 * 1 ST
Product Details & Documents

Borosilicate glass 5,4, amber.

  • Interchangeable key
  • With glass or PTFE key
  • Key with retention device

Stopcock housing with burette tip and key.

Specifications
  • Description:
    Spare PTFE key for 25 ml and 50 ml burette capacity
  • Seq:
    08
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.