Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Sampling siphons

Sampling siphons

Sampling siphon, PE, 275 ml, 1000 mm
Catalog # BURK5387-0100
Supplier:   BURKLE
Sampling siphons
Catalog # BURK5387-0100
Supplier:  BURKLE
Sampling siphons
Sampling siphons

Sampling siphon, PE, 275 ml, 1000 mm

Price
SFr. 40.40
Log in for availability
1 * 1 ST
Product Details & Documents

Disposable plunging samplers acting like siphons.

  • Transparent plastic for better visibility
  • PE surface resists adhesion of the material
  • Minimised sample retention on the dip sampler

If strongly adhesive liquids are sampled, it is easier to dispose of the sampler than to carry out a complicated cleaning process.

Specifications
  • Capacity:
    275 ml
  • Description:
    Sampling siphon
  • Length:
    1000 mm
  • Material:
    PE
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.