Orders LinkContactUsLinkComponent
Membrane Centramate™ cassettes
Membrane Centramate™ cassettes
0,65 µm, 200 cm²
Catalog # 516-9715
Membrane Centramate™ cassettes
Catalog # 516-9715
Membrane Centramate™ cassettes
Membrane Centramate™ cassettes
0,65 µm, 200 cm²
Price
SFr. 622.00
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Filtration area:
    200 cm²
  • Pore size:
    0,65 µm
  • Seq:
    14