Orders LinkContactUsLinkComponent
Pycnometers, unadjusted
Pycnometers, unadjusted
25 ml
Catalog # 613-2176
Supplier:   witeg Labortechnik
Pycnometers, unadjusted
Catalog # 613-2176
Supplier:  witeg Labortechnik
Pycnometers, unadjusted
Pycnometers, unadjusted
25 ml
Price
SFr. 35.10
Log in for availability
1 * 1 ST
Product Details & Documents
Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.
  • With ground-in capillary stopper
  • Nominal capacity printed on the bottom
  • Very good chemical and temperature resistance
 : DIN 12797
Specifications
  • Volume (nom.):
    25 ml