Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Transfer pipettes

Ungraduated, 3,0 ml, 33 µl, 150 mm, -, Bulk
  612-1755
 :   RATIOLAB
Transfer pipettes
  612-1755
 :  RATIOLAB
Transfer pipettes
Transfer pipettes

Ungraduated, 3,0 ml, 33 µl, 150 mm, -, Bulk

 
SFr. 34.90
   
1 * 500 ST
 

PE, disposable.

  • Soft, elastic bulb
  • Unbreakable, reduced risk of injury compared to glass
  • Inert to biological fluids and most acids
  • Graduated pipettes with clearly visible embossed graduations
  • Sterile pipettes are gas-sterilised and individually packed
  • Suitable for food contact
 
  • Packed:
    Bulk
  • Sterile:
    -
  • Capacity:
    3,0 ml
  • Description:
    Ungraduated
  • Length:
    150 mm
  • Seq:
    15
  • Drop vol.:
    33 µl