Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Transfer pipettes

Ungraduated, 0,2 ml, 30 µl, 130 mm, -, Bulk
Catalog # 612-1732
Supplier:   RATIOLAB
Transfer pipettes
Catalog # 612-1732
Supplier:  RATIOLAB
Transfer pipettes
Transfer pipettes

Ungraduated, 0,2 ml, 30 µl, 130 mm, -, Bulk

Price
SFr. 88.80
Log in for availability
1 * 500 ST
Product Details & Documents

PE, disposable.

  • Soft, elastic bulb
  • Unbreakable, reduced risk of injury compared to glass
  • Inert to biological fluids and most acids
  • Graduated pipettes with clearly visible embossed graduations
  • Sterile pipettes are gas-sterilised and individually packed
  • Suitable for food contact
Specifications
  • Packed:
    Bulk
  • Sterile:
    -
  • Capacity:
    0,2 ml
  • Description:
    Ungraduated
  • Length:
    130 mm
  • Seq:
    13
  • Drop vol.:
    30 µl