Orders LinkContactUsLinkComponent
Burners
Burners
Bunsen burner with stopcock, Natural gas
Catalog # 451-1330
Supplier:   USBECK Laborgeräte
Burners
Catalog # 451-1330
Burners
Burners
Bunsen burner with stopcock, Natural gas
Price
SFr. 77.00
Log in for availability
1 * 1 ST
Product Details & Documents
DIN type.
  • Air regulation and pilot flame inside
Ordering information: Supplied without safety base, this needs to be ordered separately.
Specifications
  • Description:
    Bunsen burner with stopcock
  • For:
    Natural gas