Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Tissue grinders

Tissue grinders

Plunger with PTFE head and stainless steel shaft, for 5 mm tubes, 235 mm
Catalog # 432-1240
Supplier:   DELTALAB
Tissue grinders
Catalog # 432-1240
Supplier:  DELTALAB
Tissue grinders

Plunger with PTFE head and stainless steel shaft, for 5 mm tubes, 235 mm

Price
SFr. 79.30
Log in for availability
1 * 1 ST
Product Details & Documents

Clearance between Glass vessel and appropriately sized plunger is approximately 200 µm.

Specifications
  • Description:
    Plunger with PTFE head and stainless steel shaft, for 5 mm tubes
  • Seq:
    06
  • Total length:
    235 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.