Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Tissue grinders

Tissue grinders

Plunger with PTFE head and stainless steel shaft, for 2 mm tubes, 230 mm
Catalog # 432-1235
Supplier:   DELTALAB
Tissue grinders
Catalog # 432-1235
Supplier:  DELTALAB
Tissue grinders

Plunger with PTFE head and stainless steel shaft, for 2 mm tubes, 230 mm

Price
SFr. 81.40
Log in for availability
1 * 1 ST
Product Details & Documents

Clearance between Glass vessel and appropriately sized plunger is approximately 200 µm.

Specifications
  • Description:
    Plunger with PTFE head and stainless steel shaft, for 2 mm tubes
  • Seq:
    05
  • Total length:
    230 mm
Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..